Gene Transcripts
- Gene Symbol
- C1qtnf1
- Entrez Gene ID
- 56745
- Gene Name
- C1q and tumor necrosis factor related protein 1
- Gene Aliases
- 1600017K21Rik, CTRP1, Zsig37
- Chromosome Location
- Chr. 11: 118428457 - 118451782 on Build GRCm38
- UniGene ID
- Mm.23845
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001204129.1 |
NP_001191058.1
|
2, 3 |
279 |
|
NM_001204130.1 |
NP_001191059.1
|
2, 3 |
191 |
|
NM_019959.3 |
NP_064343.1
|
2, 3 |
269 |
|
GenBank mRNA |
AF192499.1 |
- |
2, 3 |
217 |
|
AK005484.1 |
- |
2, 3 |
238 |
|
AK145104.1 |
- |
2, 3 |
192 |
|
BC037636.1 |
- |
1, 2 |
184 |
|
BC049894.1 |
- |
2, 3 |
220 |
4018549 |
CJ056741.1 |
- |
2, 3 |
269 |
|
DQ002394.1 |
- |
1, 2 |
163 |
|
View Full Product Details