Gene Transcripts

Gene Symbol
Caln1
Entrez Gene ID
140904
Gene Name
calneuron 1
Gene Aliases
9630012C17Rik, Cabp8, MNCb-0849
Chromosome Location
Chr. 5: 130369356 - 130846325 on Build GRCm38
UniGene ID
Mm.320206
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021371.2 NP_067346.1 4 955
NM_181045.1 NP_851388.1 5 655
GenBank mRNA AB041539.1 - 4 946
AF282251.1 - 5 637
AK035865.1 - 5 624
AK081011.1 - 4 389
AK158641.1 - 4 317
AK158892.1 - 4 927
AK163648.1 - 4 418
BC057129.1 - 4 934 6811387
BC058963.1 - 4 934 6811387

View Full Product Details