Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022992.2 | NP_075368.1 | 3 | 1202 | |
GenBank mRNA | AF421860.1 | - | 3 | 1209 | |
AK005259.1 | - | 3 | 1198 | ||
AK008519.1 | - | 3 | 1182 | ||
AK076519.1 | - | 3 | 1199 | ||
AK077826.1 | - | 3 | 1208 | ||
AK088205.1 | - | 3 | 1197 | ||
BC003897.1 | - | 3 | 1202 | 3600236 | |
D87211.1 | - | 3 | 1245 |