Gene Transcripts

Gene Symbol
Anapc4
Entrez Gene ID
52206
Gene Name
anaphase promoting complex subunit 4
Gene Aliases
2610306D21Rik, APC4, D5Ertd249e
Chromosome Location
Chr. 5: 52834126 - 52866742 on Build GRCm38
UniGene ID
Mm.272568
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024213.2 NP_077175.1 2 330
GenBank mRNA AK088759.1 - 2 320
AK146000.1 - 2 327
AK148625.1 - 2 339
AK166061.1 - 2 453
AK168031.1 - 2 327
AK168897.1 - 2 339
BC016237.1 - 2 313 4504779
BC024870.1 - 2 330 4949079

View Full Product Details