Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026330.3 | NP_080606.2 | 5 | 561 | |
GenBank mRNA | AK009715.1 | - | 5 | 450 | |
AK011011.1 | - | 5 | 466 | ||
AK160217.1 | - | 5 | 461 | ||
AK166410.1 | - | 5 | 454 | ||
BC031848.1 | - | 5 | 540 | 3984224 | |
BC049558.1 | - | 5 | 554 | 6707952 |