Gene Transcripts
- Gene Symbol
- Arpc4
- Entrez Gene ID
- 68089
- Gene Name
- actin related protein 2/3 complex, subunit 4
- Gene Aliases
- 20kDa, 5330419I20Rik, AI327076, p20-Arc
- Chromosome Location
- Chr. 6: 113378113 - 113390447 on Build GRCm38
- UniGene ID
- Mm.289306
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026552.3 |
NP_080828.1
|
2, 3 |
223 |
|
GenBank mRNA |
AJ278129.1 |
- |
2, 3 |
149 |
|
AK017993.1 |
- |
2, 3 |
224 |
|
AK018069.1 |
- |
2, 3 |
161 |
|
AK039215.1 |
- |
2, 3 |
171 |
|
AK076248.1 |
- |
2, 3 |
171 |
|
AK077325.1 |
- |
2, 3 |
145 |
|
AK133626.1 |
- |
2, 3 |
171 |
|
AK136082.1 |
- |
2, 3 |
166 |
|
AK150690.1 |
- |
2, 3 |
157 |
|
AK153380.1 |
- |
2, 3 |
155 |
|
AK166440.1 |
- |
2, 3 |
145 |
|
AK193553.1 |
- |
1, 2 |
94 |
|
AK194624.1 |
- |
1, 2 |
94 |
|
AK194784.1 |
- |
1, 2 |
94 |
|
AK197517.1 |
- |
1, 2 |
94 |
|
BC015280.1 |
- |
1, 2 |
135 |
|
BC055309.1 |
- |
1, 2 |
134 |
6529255 |
View Full Product Details