Gene Transcripts

Gene Symbol
Eqtn
Entrez Gene ID
67753
Gene Name
equatorin, sperm acrosome associated
Gene Aliases
1700028B15Rik, 4930579C15Rik, Afaf, C81058, Eqtl, Eqts
Chromosome Location
Chr. 4: 94875696 - 94929950 on Build GRCm38
UniGene ID
Mm.25097
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290623.1 NP_001277552.1 3 361
NM_027089.4 NP_081365.2 3 361
GenBank mRNA AB438105.1 - 3 270
AB438106.1 - 3 270
AK006446.1 - 3 312
AK016313.1 - 3 329
AK016738.1 - 3 332
BC089517.1 - 3 328 6740450
BY716419.1 - 3 328
DQ336137.1 - 3 312
EU828781.1 - 3 346
EU828782.1 - 3 346

View Full Product Details