Gene Transcripts

Gene Symbol
Acoxl
Entrez Gene ID
74121
Gene Name
acyl-Coenzyme A oxidase-like
Gene Aliases
1200014P05Rik, AV025673
Chromosome Location
Chr. 2: 127838797 - 128126252 on Build GRCm38
UniGene ID
Mm.244835
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028765.3 NP_083041.1 1 298
GenBank mRNA AK004775.1 - 1 300

View Full Product Details