Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000382.2 | NP_000373.1 | 1 | 346 | |
NM_001031806.1 | NP_001026976.1 | 1 | 346 | ||
XM_011523732.1 | XP_011522034.1 | 2 | 255 | ||
XM_011523733.1 | XP_011522035.1 | 2 | 292 | ||
XM_017024355.1 | XP_016879844.1 | 1 | 148 | ||
XM_017024356.1 | XP_016879845.1 | 2 | 254 | ||
XM_017024357.1 | XP_016879846.1 | 2 | 255 | ||
XM_017024358.1 | XP_016879847.1 | 2 | 292 | ||
GenBank mRNA | AB208894.1 | - | 1 | 54 | |
AK025677.1 | - | 1 | 246 | ||
AK292381.1 | - | 1 | 313 | ||
AK315096.1 | - | 1 | 175 | ||
BC002430.2 | - | 1 | 166 | 3346862 | |
CR457422.1 | - | 1 | 125 | ||
L47162.1 | - | 1 | 288 | ||
U46689.1 | - | 1 | 574 |