Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029682.4 | NP_083958.3 | 9 | 1539 | |
GenBank mRNA | AB010121.1 | - | 9 | 1234 | |
AB010122.1 | - | 9 | 1198 | ||
AB066211.1 | - | 9 | 1539 | ||
AB066212.1 | - | 5 | 888 | ||
AB066213.1 | - | 9 | 1222 | ||
AK007087.1 | - | 5 | 872 | ||
BC018343.1 | - | 9 | 1265 | 4010423 | |
BC023097.1 | - | 9 | 1508 |