Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030709.2 | NP_109634.2 | 2, 3 | 191 | |
GenBank mRNA | AB016229.1 | - | 2, 3 | 199 | |
AB016230.1 | - | 2, 3 | 191 | ||
AB041037.1 | - | 2, 3 | 148 | ||
AK029714.1 | - | 3, 4 | 1038 | ||
BC132151.1 | - | 1, 2 | 137 | 40130428 | |
BC145027.1 | - | 1, 2 | 137 | 9053575 |