Gene Transcripts

Gene Symbol
Actn2
Entrez Gene ID
11472
Gene Name
actinin alpha 2
Gene Aliases
1110008F24Rik
Chromosome Location
Chr. 13: 12269426 - 12340732 on Build GRCm38
UniGene ID
Mm.37638
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033268.4 NP_150371.4 9 1082
GenBank mRNA AF248643.1 - 9 850
AY036877.1 - 10 1026
BC089579.1 - 9 1082 6757718

View Full Product Details