Gene Transcripts
- Gene Symbol
- Rnf144a
- Entrez Gene ID
- 108089
- Gene Name
- ring finger protein 144A
- Gene Aliases
- Rnf144, UIP4, Ubce7ip4
- Chromosome Location
- Chr. 12: 26306794 - 26415321 on Build GRCm38
- UniGene ID
- Mm.214932
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001081977.2 |
NP_001075446.1
|
5, 6 |
668 |
|
NM_080563.4 |
NP_542130.1
|
4, 5 |
594 |
|
GenBank mRNA |
AF360998.1 |
- |
5, 6 |
664 |
|
AK038586.1 |
- |
4, 5 |
486 |
|
AK133544.1 |
- |
2, 3 |
208 |
|
AK172904.1 |
- |
3, 4 |
429 |
|
BC030187.1 |
- |
4, 5 |
558 |
5345434 |
BQ884505.1 |
- |
4, 5 |
527 |
|
BU055293.1 |
- |
6, 7 |
605 |
|
BY753828.1 |
- |
4, 5 |
587 |
|
View Full Product Details