Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163505.1 | NP_001156977.1 | 2 | 368 | |
NM_146091.4 | NP_666203.3 | 2 | 270 | ||
GenBank mRNA | AK028842.1 | - | 2 | 131 | |
AK030660.1 | - | 2 | 130 | ||
AK077752.1 | - | 2 | 133 | ||
AK147720.1 | - | 2 | 228 | ||
AK169476.1 | - | 2 | 130 | ||
AK170550.1 | - | 2 | 174 | ||
BC017138.1 | - | 2 | 119 | 4485115 | |
BY108453.1 | - | 2 | 268 | ||
CA318221.1 | - | 2 | 270 |