Gene Transcripts

Gene Symbol
C8a
Entrez Gene ID
230558
Gene Name
complement component 8, alpha polypeptide
Gene Aliases
Chromosome Location
Chr. 4: 104815679 - 104876486 on Build GRCm38
UniGene ID
Mm.197638
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290645.1 NP_001277574.1 2 228
NM_146148.2 NP_666260.1 3 325
GenBank mRNA AB077295.1 - 3 339
AB077296.1 - 3 288
AB077297.1 - 2 194
AB077298.1 - 3 288
AK090245.1 - 3 323
BC027748.1 - 3 321 5052412

View Full Product Details