Gene Transcripts
- Gene Symbol
- ACYP1
- Entrez Gene ID
- 97
- Gene Name
- acylphosphatase 1
- Gene Aliases
- ACYPE
- Chromosome Location
- Chr. 14: 75053221 - 75069483 on Build GRCh38
- UniGene ID
- Hs.18573
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001107.4 |
NP_001098.1
|
2, 3 |
196 |
|
NM_001302616.1 |
NP_001289545.1
|
2, 3 |
178 |
|
NM_001302617.1 |
NP_001289546.1
|
2, 3 |
374 |
|
NR_126393.1 |
|
3, 4 |
275 |
|
NR_126394.1 |
|
1, 2 |
282 |
|
GenBank mRNA |
AK312101.1 |
- |
2, 3 |
196 |
|
BC035568.1 |
- |
2, 3 |
181 |
5392248 |
BG614847.1 |
- |
3, 4 |
242 |
|
BP316920.1 |
- |
2, 3 |
159 |
|
CD675334.1 |
- |
2, 3 |
383 |
|
DB071115.1 |
- |
2, 3 |
196 |
|
HY012032.1 |
- |
1, 2 |
244 |
|
X84194.1 |
- |
1, 2 |
160 |
|
View Full Product Details