Gene Transcripts

Gene Symbol
Hps6
Entrez Gene ID
20170
Gene Name
Hermansky-Pudlak syndrome 6
Gene Aliases
5330434M19Rik, BLOC2, ru
Chromosome Location
Chr. 19: 46003478 - 46006173 on Build GRCm38
UniGene ID
Mm.101110
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176785.3 NP_789742.2 1 1213
GenBank mRNA AF536239.1 - 1 1158
AK030582.1 - 1 1184
AK040849.1 - 1 1160
AK159588.1 - 1 1179

View Full Product Details