Gene Transcripts

Gene Symbol
Btla
Entrez Gene ID
208154
Gene Name
B and T lymphocyte associated
Gene Aliases
A630002H24
Chromosome Location
Chr. 16: 45223547 - 45252895 on Build GRCm38
UniGene ID
Mm.38199
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037719.2 NP_001032808.2 1, 2 128
NM_177584.3 NP_808252.1 1, 2 128
GenBank mRNA AK041334.1 - 1, 2 130

View Full Product Details