Gene Transcripts

Gene Symbol
Ppm1n
Entrez Gene ID
232941
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent, 1N (putative)
Gene Aliases
A930002H20, C79127
Chromosome Location
Chr. 7: 19276807 - 19280194 on Build GRCm38
UniGene ID
Mm.134068
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177691.3 NP_808359.1 5 1391
GenBank mRNA AK044232.1 - 5 1391
AK044334.1 - 5 1392
BC096372.1 - 5 1380 4507828

View Full Product Details