Gene Transcripts

Gene Symbol
ASTN1
Entrez Gene ID
460
Gene Name
astrotactin 1
Gene Aliases
ASTN
Chromosome Location
Chr. 1: 176855118 - 177164904 on Build GRCh38
UniGene ID
Hs.495897
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286164.1 NP_001273093.1 11 2008
NM_004319.2 NP_004310.1 11 2008
NM_207108.2 NP_996991.1 11 2008
XM_017001340.1 XP_016856829.1 11 1982
XM_017001341.1 XP_016856830.1 11 1982
GenBank mRNA AB006627.1 - 11 1806
AK295126.1 - 11 1960
AK307820.1 - 11 1918
AK309094.1 - 11 2077
BC142697.1 - 11 1787 40148746
BC146795.1 - 11 1806 9007266

View Full Product Details