Gene Transcripts

Gene Symbol
BRDT
Entrez Gene ID
676
Gene Name
bromodomain testis associated
Gene Aliases
BRD6, CT9
Chromosome Location
Chr. 1: 91949371 - 92014428 on Build GRCh38
UniGene ID
Hs.482520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242805.2 NP_001229734.2 9 1682
NM_001242806.2 NP_001229735.2 8 1625
NM_001242807.2 NP_001229736.2 7 1281
NM_001242808.2 NP_001229737.2 7 1475
NM_001242810.2 NP_001229739.2 8 1969
NM_001726.4 NP_001717.3 8 1408
NM_207189.3 NP_997072.2 8 1613
XM_006710853.3 XP_006710916.1 9 2198
XM_006710854.3 XP_006710917.1 10 2043
XM_006710855.3 XP_006710918.1 10 2048
XM_006710856.3 XP_006710919.1 10 2022
XM_006710857.3 XP_006710920.1 10 2012
XM_011542032.2 XP_011540334.1 10 1532
XM_011542033.2 XP_011540335.1 10 1506
XM_011542034.2 XP_011540336.1 10 1522
XM_011542035.2 XP_011540337.1 10 1496
XM_011542036.2 XP_011540338.1 8 1411
GenBank mRNA AF019085.1 - 8 1371
AK302758.1 - 7 1217
AK303008.1 - 8 1373
AK316442.1 - 7 1280
AY338951.1 - 9 1682
BC005281.1 - 8 1339
BC017582.1 - 8 1414
BC047900.1 - 8 1378
BC062700.1 - 8 1373
BX648135.1 - 8 1704

View Full Product Details