Gene Transcripts

Gene Symbol
BUB1
Entrez Gene ID
699
Gene Name
BUB1 mitotic checkpoint serine/threonine kinase
Gene Aliases
BUB1A, BUB1L, hBUB1
Chromosome Location
Chr. 2: 110637698 - 110678107 on Build GRCh38
UniGene ID
Hs.469649
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278616.1 NP_001265545.1 21 2780
NM_004336.4 NP_004327.1 22 2840
GenBank mRNA AF011387.1 - 14 1905
AF043294.2 - 22 2760
AF046078.1 - 22 2777
AF047471.1 - 22 2763
AF053305.1 - 22 2791
AI635394.1 - 4 663
AK291221.1 - 22 2793
AK302418.1 - 21 2736
BC028201.1 - 22 2761 5205385

View Full Product Details