Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006888.4 | NP_008819.1 | 3 | 337 | |
XM_006720258.3 | XP_006720321.1 | 3 | 966 | ||
GenBank mRNA | AK289937.1 | - | 4 | 284 | |
AU122722.1 | - | 3 | 291 | ||
AY189287.1 | - | 2 | 2218 | ||
BC000454.2 | - | 3 | 271 | 2821489 | |
BC007965.2 | - | 3 | 313 | 4308380 | |
BC008597.1 | - | 3 | 271 | 3452971 | |
BC011834.2 | - | 3 | 291 | 3942245 | |
BC047523.1 | - | 3 | 291 | ||
BT006818.1 | - | 2 | 89 | ||
M27319.1 | - | 3 | 176 |