Gene Transcripts

Gene Symbol
RTCA
Entrez Gene ID
8634
Gene Name
RNA 3'-terminal phosphate cyclase
Gene Aliases
RPC, RTC1, RTCD1
Chromosome Location
Chr. 1: 100266158 - 100292769 on Build GRCh38
UniGene ID
Hs.484222
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130841.1 NP_001124313.1 12 1303
NM_003729.3 NP_003720.1 11 1264
XM_005271298.4 XP_005271355.1 11 1261
XM_005271299.4 XP_005271356.1 11 1250
XM_017002669.1 XP_016858158.1 11 1007
GenBank mRNA AK292366.1 - 11 1213
AK307445.1 - 9 1000
BC012604.1 - 12 1217 4281547
Y11651.1 - 11 1216

View Full Product Details