Gene Transcripts

Gene Symbol
AP3D1
Entrez Gene ID
8943
Gene Name
adaptor related protein complex 3 delta 1 subunit
Gene Aliases
ADTD, hBLVR
Chromosome Location
Chr. 19: 2100988 - 2151620 on Build GRCh38
UniGene ID
Hs.512815
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001261826.1 NP_001248755.1 2, 3 403
NM_003938.6 NP_003929.4 2, 3 403
XM_006722932.2 XP_006722995.1 2, 3 466
GenBank mRNA AB208804.1 - 2, 3 387
AF002163.1 - 491
BC005142.2 - 2, 3 231
BC010065.2 - 2, 3 382 4106961
BQ227203.1 - 2, 3 403
U91930.1 - 2, 3 389

View Full Product Details