Gene Transcripts

Gene Symbol
HSPA4L
Entrez Gene ID
22824
Gene Name
heat shock protein family A (Hsp70) member 4 like
Gene Aliases
APG-1, HSPH3, Osp94
Chromosome Location
Chr. 4: 127781821 - 127834073 on Build GRCh38
UniGene ID
Hs.135554
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317381.1 NP_001304310.1 12 1653
NM_001317382.1 NP_001304311.1 11 1372
NM_001317383.1 NP_001304312.1 10 1540
NM_014278.3 NP_055093.2 11 1663
XM_011531745.2 XP_011530047.1 11 1732
GenBank mRNA AB023421.1 - 11 1375
AK027822.1 - 11 1550
AK302115.1 - 11 1346
AK303053.1 - 12 1653
AK307464.1 - 10 1427
AY290862.1 - 11 1529
BC040560.1 - 11 1574 5270824
EF197155.1 - 11 1321

View Full Product Details