Gene Transcripts

Gene Symbol
ASF1A
Entrez Gene ID
25842
Gene Name
anti-silencing function 1A histone chaperone
Gene Aliases
CGI-98, CIA, HSPC146
Chromosome Location
Chr. 6: 118894077 - 118909171 on Build GRCh38
UniGene ID
Hs.292316
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014034.2 NP_054753.1 3 696
GenBank mRNA AB028628.1 - 3 522
AF151856.1 - 5 973
AF161495.1 - 5 973
AF279306.1 - 3 508
AK025738.1 - 3 553
AL050261.1 - 3 551
BC010878.1 - 3 535 3546858
CR457347.1 - 3 359

View Full Product Details