Gene Transcripts

Gene Symbol
ACAD9
Entrez Gene ID
28976
Gene Name
acyl-CoA dehydrogenase family member 9
Gene Aliases
NPD002
Chromosome Location
Chr. 3: 128879490 - 128916075 on Build GRCh38
UniGene ID
Hs.567482
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014049.4 NP_054768.2 7, 8 1012
NR_033426.1 7, 8 1188
XM_017006264.1 XP_016861753.1 7, 8 1176
XM_017006265.1 XP_016861754.1 1, 2 158
GenBank mRNA AB209427.1 - 7, 8 2705
AF327351.1 - 7, 8 842
AK022568.1 - 6, 7 938
AK024012.1 - 7, 8 856
AK130311.1 - 7, 8 883
AK294597.1 - 7, 8 680
AK297952.1 - 7, 8 1056
AK310644.1 - 6, 7 785
BC007970.2 - 7, 8 889 4304209
BC013354.2 - 7, 8 866 3935925

View Full Product Details