Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016249.3 | NP_057333.1 | 3 | 407 | |
GenBank mRNA | AF116194.1 | - | 3 | 390 | |
AF151378.1 | - | 3 | 388 | ||
AF196482.1 | - | 3 | 388 | ||
AK313591.1 | - | 3 | 400 | ||
BC005891.2 | - | 1 | 138 | 3050294 | |
BC013318.1 | - | 3 | 411 | 4133159 | |
CR457168.1 | - | 1 | 59 |