Gene Transcripts
- Gene Symbol
- APBB1IP
- Entrez Gene ID
- 54518
- Gene Name
- amyloid beta precursor protein binding family B member 1 interacting protein
- Gene Aliases
- INAG1, PREL1, RARP1, RIAM
- Chromosome Location
- Chr. 10: 26438324 - 26567879 on Build GRCh38
- UniGene ID
- Hs.310421
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019043.3 |
NP_061916.3
|
3, 4 |
391 |
|
XM_006717451.2 |
XP_006717514.1
|
2, 3 |
89 |
|
XM_011519514.2 |
XP_011517816.1
|
3, 4 |
404 |
|
GenBank mRNA |
AB085852.1 |
- |
1, 2 |
70 |
|
AK000812.1 |
- |
3, 4 |
350 |
|
AK303597.1 |
- |
3, 4 |
951 |
|
AK308916.1 |
- |
3, 4 |
372 |
|
AY152730.1 |
- |
2, 3 |
157 |
|
BC035636.1 |
- |
3, 4 |
303 |
|
BC054516.1 |
- |
2, 3 |
169 |
5172946 |
BM150270.1 |
- |
3, 4 |
392 |
|
BM556115.1 |
- |
|
358 |
|
View Full Product Details