Gene Transcripts
- Gene Symbol
- AHI1
- Entrez Gene ID
- 54806
- Gene Name
- Abelson helper integration site 1
- Gene Aliases
- AHI-1, JBTS3, ORF1, dJ71N10.1
- Chromosome Location
- Chr. 6: 135283972 - 135497775 on Build GRCh38
- UniGene ID
- Hs.386684
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001134830.1 |
NP_001128302.1
|
17, 18 |
2867 |
|
NM_001134831.1 |
NP_001128303.1
|
19, 20 |
3014 |
|
NM_001134832.1 |
NP_001128304.1
|
18, 19 |
2929 |
|
NM_017651.4 |
NP_060121.3
|
18, 19 |
2929 |
|
XM_011535910.2 |
XP_011534212.1
|
20, 21 |
3158 |
|
XM_011535911.2 |
XP_011534213.1
|
18, 19 |
2888 |
|
XM_017010977.1 |
XP_016866466.1
|
19, 20 |
3133 |
|
XM_017010978.1 |
XP_016866467.1
|
18, 19 |
2831 |
|
XM_017010979.1 |
XP_016866468.1
|
18, 19 |
2831 |
|
XM_017010980.1 |
XP_016866469.1
|
18, 19 |
2831 |
|
XM_017010981.1 |
XP_016866470.1
|
18, 19 |
2966 |
|
XM_017010982.1 |
XP_016866471.1
|
18, 19 |
2831 |
|
XM_017010983.1 |
XP_016866472.1
|
18, 19 |
2831 |
|
XM_017010984.1 |
XP_016866473.1
|
18, 19 |
2831 |
|
GenBank mRNA |
AJ459824.1 |
- |
18, 19 |
2909 |
|
AJ459825.1 |
- |
18, 19 |
2909 |
|
AJ606362.1 |
- |
17, 18 |
2847 |
|
AK000076.1 |
- |
10, 11 |
1330 |
|
AK024085.1 |
- |
6, 7 |
822 |
|
AK092262.1 |
- |
17, 18 |
2840 |
|
AK225210.1 |
- |
10, 11 |
1330 |
|
AL136797.1 |
- |
16, 17 |
2648 |
|
BC015019.1 |
- |
8, 9 |
1128 |
|
BC094800.1 |
- |
19, 20 |
2937 |
4823262 |
DQ090887.1 |
- |
19, 20 |
2968 |
|
View Full Product Details