Gene Transcripts
- Gene Symbol
- ARHGEF40
- Entrez Gene ID
- 55701
- Gene Name
- Rho guanine nucleotide exchange factor 40
- Gene Aliases
- SOLO
- Chromosome Location
- Chr. 14: 21061276 - 21090248 on Build GRCh38
- UniGene ID
- Hs.35125
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278529.1 |
NP_001265458.1
|
5, 6 |
1874 |
|
NM_001278530.1 |
NP_001265459.1
|
5, 6 |
1707 |
|
NM_018071.4 |
NP_060541.3
|
5, 6 |
1874 |
|
XM_005267844.2 |
XP_005267901.1
|
5, 6 |
1910 |
|
XM_011536937.2 |
XP_011535239.1
|
5, 6 |
1910 |
|
XM_011536938.2 |
XP_011535240.1
|
5, 6 |
1912 |
|
XM_017021434.1 |
XP_016876923.1
|
6, 7 |
2167 |
|
GenBank mRNA |
AK024463.1 |
- |
3, 4 |
1112 |
|
AK074057.1 |
- |
5, 6 |
1819 |
|
BC142692.1 |
- |
5, 6 |
1817 |
40148708 |
BC151212.1 |
- |
5, 6 |
1817 |
|
View Full Product Details