Gene Transcripts

Gene Symbol
ADGRG6
Entrez Gene ID
57211
Gene Name
adhesion G protein-coupled receptor G6
Gene Aliases
APG1, DREG, GPR126, LCCS9, PS1TP2, VIGR
Chromosome Location
Chr. 6: 142301919 - 142446266 on Build GRCh38
UniGene ID
Hs.743302
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001032394.2 NP_001027566.1 10 1929
NM_001032395.2 NP_001027567.1 10 1929
NM_020455.5 NP_065188.4 11 2013
NM_198569.2 NP_940971.1 11 2013
XM_005267061.3 XP_005267118.1 11 1914
XM_006715516.2 XP_006715579.1 11 1914
XM_006715517.2 XP_006715580.1 11 1714
XM_006715518.2 XP_006715581.1 10 1830
XM_011535964.1 XP_011534266.1 11 1717
XM_017011085.1 XP_016866574.1 11 1914
GenBank mRNA AB183546.1 - 11 2003
AB183547.1 - 10 1919
AB183548.1 - 11 2003
AB183549.1 - 10 1919
AF216967.1 - 10 1934
AK307543.1 - 10 1827
BC075798.1 - 10 1989 30333185
BX640873.1 - 10 1535
BX640971.1 - 11 1939
BX648315.1 - 10 1642

View Full Product Details