Gene Transcripts

Gene Symbol
AFAP1
Entrez Gene ID
60312
Gene Name
actin filament associated protein 1
Gene Aliases
AFAP, AFAP-110, AFAP110
Chromosome Location
Chr. 4: 7758713 - 7939926 on Build GRCh38
UniGene ID
Hs.529369
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134647.1 NP_001128119.1 9 1254
NM_198595.2 NP_940997.1 9 1254
XM_006713909.3 XP_006713972.1 9 1137
XM_011513544.2 XP_011511846.1 9 1307
XM_017008535.1 XP_016864024.1 8 983
XM_017008536.1 XP_016864025.1 8 983
GenBank mRNA AB209676.1 - 6 707
AF188700.1 - 8 981
AK290807.1 - 9 1081
AK297631.1 - 9 1047
BC032777.1 - 8 1005 4914388

View Full Product Details