Gene Transcripts
- Gene Symbol
- TMEM243
- Entrez Gene ID
- 79161
- Gene Name
- transmembrane protein 243
- Gene Aliases
- C7orf23, MM-TRAG, MMTRAG
- Chromosome Location
- Chr. 7: 87195593 - 87220603 on Build GRCh38
- UniGene ID
- Hs.619593
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_005250586.3 |
XP_005250643.1
|
2, 3 |
509 |
|
XM_005250587.4 |
XP_005250644.1
|
2, 3 |
643 |
|
XM_005250588.3 |
XP_005250645.1
|
2, 3 |
341 |
|
XM_005250589.3 |
XP_005250646.1
|
2, 3 |
509 |
|
XM_017012616.1 |
XP_016868105.1
|
1, 2 |
930 |
|
XM_017012617.1 |
XP_016868106.1
|
2, 3 |
632 |
|
XM_017012618.1 |
XP_016868107.1
|
2, 3 |
376 |
|
GenBank mRNA |
AK313794.1 |
- |
1, 2 |
311 |
|
AL711214.1 |
- |
2, 3 |
347 |
|
BC002837.2 |
- |
1, 2 |
293 |
3634983 |
BP236768.1 |
- |
2, 3 |
400 |
|
BT006973.1 |
- |
1, 2 |
81 |
|
CB993361.1 |
- |
2, 3 |
550 |
|
View Full Product Details