Gene Transcripts

Gene Symbol
OR4P4
Entrez Gene ID
81300
Gene Name
olfactory receptor family 4 subfamily P member 4
Gene Aliases
OR4P3P
Chromosome Location
Chr. 11: 55638358 - 55639296 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004124.2 NP_001004124.1 1 326

View Full Product Details