Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001077484.1 | NP_001070952.1 | 13 | 1728 | |
NM_001278387.1 | NP_001265316.1 | 14 | 2190 | ||
NM_001278388.1 | NP_001265317.1 | 12 | 1952 | ||
NM_001278389.1 | NP_001265318.1 | 13 | 1217 | ||
NM_001278390.1 | NP_001265319.1 | 13 | 1930 | ||
NM_030674.3 | NP_109599.3 | 13 | 2067 | ||
XM_011538784.2 | XP_011537086.1 | 13 | 1452 | ||
XM_011538786.2 | XP_011537088.1 | 13 | 1452 | ||
XM_011538787.2 | XP_011537089.1 | 13 | 1452 | ||
XM_017019990.1 | XP_016875479.1 | 13 | 1682 | ||
GenBank mRNA | AF247166.1 | - | 13 | 1413 | |
AF271070.1 | - | 13 | 1311 | ||
AK027825.1 | - | 13 | 1217 | ||
AK074758.1 | - | 6 | 484 | ||
AK074797.1 | - | 10 | 786 | ||
AK074851.1 | - | 11 | 929 | ||
AK074949.1 | - | 11 | 975 | ||
AK130314.1 | - | 1 | 1211 | ||
AK225639.1 | - | 13 | 1305 | ||
AK225778.1 | - | 13 | 1327 | ||
AK308129.1 | - | 11 | 1015 | ||
AK309410.1 | - | 13 | 1644 | ||
AK313731.1 | - | 11 | 1018 | ||
BC010620.1 | - | 13 | 1274 | 3871101 | |
BX647547.1 | - | 14 | 1762 | ||
BX648495.1 | - | 13 | 1655 | ||
BX648676.1 | - | 12 | 1522 |