Gene Transcripts

Gene Symbol
PDCD2L
Entrez Gene ID
84306
Gene Name
programmed cell death 2 like
Gene Aliases
Chromosome Location
Chr. 19: 34404398 - 34426168 on Build GRCh38
UniGene ID
Hs.515344
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032346.1 NP_115722.1 4 454
GenBank mRNA BC006146.1 - 4 454

View Full Product Details