Gene Transcripts

Gene Symbol
R3HDM4
Entrez Gene ID
91300
Gene Name
R3H domain containing 4
Gene Aliases
C19orf22
Chromosome Location
Chr. 19: 896503 - 913225 on Build GRCh38
UniGene ID
Hs.557655
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138774.3 NP_620129.2 3, 4 418
XM_011528416.2 XP_011526718.1 3, 4 409
GenBank mRNA BC012775.2 - 3, 4 393

View Full Product Details