Gene Transcripts
- Gene Symbol
- C11orf52
- Entrez Gene ID
- 91894
- Gene Name
- chromosome 11 open reading frame 52
- Gene Aliases
- Chromosome Location
- Chr. 11: 111918877 - 111926871 on Build GRCh38
- UniGene ID
- Hs.97013
- Species
- Homo sapiens
- Gene Symbol
- HSPB2-C11orf52
- Entrez Gene ID
- 100528019
- Gene Name
- HSPB2-C11orf52 readthrough (NMD candidate)
- Gene Aliases
- Chromosome Location
- Chr. 11: 111912736 - 111926871 on Build GRCh38
- UniGene ID
- Hs.97013
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037651.1 |
|
5 |
658 |
|
GenBank mRNA |
BC110872.1 |
- |
5 |
653 |
|
View Full Product Details