Gene Transcripts

Gene Symbol
CAPN13
Entrez Gene ID
92291
Gene Name
calpain 13
Gene Aliases
Chromosome Location
Chr. 2: 30610382 - 30809527 on Build GRCh38
UniGene ID
Hs.660911
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144575.2 NP_653176.2 5, 6 708
XM_011533159.2 XP_011531461.1 5, 6 705
XM_011533160.2 XP_011531462.1 6, 7 896
XM_011533161.2 XP_011531463.1 5, 6 705
XM_011533163.2 XP_011531465.1 5, 6 705
XM_017005264.1 XP_016860753.1 6, 7 896
XM_017005265.1 XP_016860754.1 5, 6 705
XM_017005266.1 XP_016860755.1 5, 6 705
GenBank mRNA AK074418.1 - 6, 7 775
AK290298.1 - 5, 6 709
AK308037.1 - 5, 6 685
BC117345.1 - 5, 6 669 40125896
BC117347.1 - 5, 6 669 40125898
BX640793.1 - 5, 6 708

View Full Product Details