Gene Transcripts
- Gene Symbol
- UBE2Q2
- Entrez Gene ID
- 92912
- Gene Name
- ubiquitin conjugating enzyme E2 Q2
- Gene Aliases
- Chromosome Location
- Chr. 15: 75843286 - 75901047 on Build GRCh38
- UniGene ID
- Hs.23033
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145335.1 |
NP_001138807.1
|
6, 7 |
945 |
|
NM_001284382.1 |
NP_001271311.1
|
5, 6 |
959 |
|
NM_173469.3 |
NP_775740.1
|
6, 7 |
1064 |
|
XM_005254787.1 |
XP_005254844.1
|
6, 7 |
836 |
|
XM_017022726.1 |
XP_016878215.1
|
6, 7 |
940 |
|
XM_017022727.1 |
XP_016878216.1
|
5, 6 |
759 |
|
GenBank mRNA |
AK000617.1 |
- |
5, 6 |
938 |
|
AK294199.1 |
- |
6, 7 |
605 |
|
AK296234.1 |
- |
6, 7 |
945 |
|
AL832429.1 |
- |
6, 7 |
889 |
|
BC006827.1 |
- |
4, 5 |
339 |
|
BC017708.1 |
- |
6, 7 |
891 |
3907760 |
View Full Product Details