Gene Transcripts

Gene Symbol
C1QTNF7
Entrez Gene ID
114905
Gene Name
C1q and tumor necrosis factor related protein 7
Gene Aliases
CTRP7, ZACRP7
Chromosome Location
Chr. 4: 15339818 - 15446169 on Build GRCh38
UniGene ID
Hs.153714
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135170.1 NP_001128642.1 2, 3 517
NM_001135171.1 NP_001128643.1 2, 3 464
NM_031911.4 NP_114117.1 2, 3 347
XM_011513772.1 XP_011512074.1 3, 4 387
XM_011513773.1 XP_011512075.1 3, 4 436
GenBank mRNA AF329839.1 - 2, 3 322
AK314802.1 - 2, 3 465
BC022187.2 - 2, 3 465 4825040
BC024015.2 - 2, 3 465 4823626
BX647781.1 - 2, 3 517
DB069980.1 - 2, 3 465
DB290077.1 - 2, 3 348

View Full Product Details