Gene Transcripts
- Gene Symbol
- C8orf34
- Entrez Gene ID
- 116328
- Gene Name
- chromosome 8 open reading frame 34
- Gene Aliases
- VEST-1, VEST1
- Chromosome Location
- Chr. 8: 68330722 - 68819023 on Build GRCh38
- UniGene ID
- Hs.491941
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001195639.1 |
NP_001182568.1
|
6, 7 |
1220 |
|
NM_052958.2 |
NP_443190.2
|
6, 7 |
1220 |
|
XM_011517445.2 |
XP_011515747.1
|
6, 7 |
1192 |
|
XM_011517446.2 |
XP_011515748.1
|
6, 7 |
1192 |
|
XM_011517448.2 |
XP_011515750.1
|
5, 6 |
1063 |
|
XM_011517449.2 |
XP_011515751.1
|
6, 7 |
1192 |
|
XM_011517450.2 |
XP_011515752.1
|
6, 7 |
1192 |
|
XM_017013019.1 |
XP_016868508.1
|
5, 6 |
1063 |
|
GenBank mRNA |
AB056652.1 |
- |
5, 6 |
2188 |
|
AK291436.1 |
- |
6, 7 |
692 |
|
AL834454.1 |
- |
6, 7 |
717 |
|
BC041961.1 |
- |
6, 7 |
1220 |
5302156 |
View Full Product Details