Gene Transcripts

Gene Symbol
ANO4
Entrez Gene ID
121601
Gene Name
anoctamin 4
Gene Aliases
TMEM16D
Chromosome Location
Chr. 12: 100717457 - 101128641 on Build GRCh38
UniGene ID
Hs.58785
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286615.1 NP_001273544.1 24 2973
NM_001286616.1 NP_001273545.1 23 2541
NM_178826.3 NP_849148.2 23 2868
XM_011537911.2 XP_011536213.1 26 2968
XM_011537912.2 XP_011536214.1 26 2968
XM_011537913.2 XP_011536215.1 25 2923
XM_011537914.2 XP_011536216.1 23 2479
XM_011537915.2 XP_011536217.1 24 2791
XM_011537916.2 XP_011536218.1 24 2632
XM_017018808.1 XP_016874297.1 23 2479
XM_017018809.1 XP_016874298.1 25 3059
XM_017018810.1 XP_016874299.1 23 2721
XM_017018811.1 XP_016874300.1 22 2676
XM_017018812.1 XP_016874301.1 22 4694
XM_017018813.1 XP_016874302.1 21 2544
XM_017018814.1 XP_016874303.1 24 4933
XM_017018815.1 XP_016874304.1 24 4933
XM_017018816.1 XP_016874305.1 24 3016
XM_017018817.1 XP_016874306.1 24 4968
GenBank mRNA AK091540.1 - 14 1489
AK091591.1 - 23 2657
AK092596.1 - 11 1055
AK293810.1 - 24 2611
AK316105.1 - 24 2612
BC109308.2 - 23 2526 40035759

View Full Product Details