Gene Transcripts

Gene Symbol
AHSA2
Entrez Gene ID
130872
Gene Name
AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast)
Gene Aliases
Hch1
Chromosome Location
Chr. 2: 61177418 - 61191210 on Build GRCh38
UniGene ID
Hs.655602
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321300.1 NP_001308229.1 5 1416
NM_001321301.1 NP_001308230.1 5 2128
NM_001321302.1 NP_001308231.1 6 836
NM_001321303.1 NP_001308232.1 4 2012
NM_001321304.1 NP_001308233.1 4 2012
NM_001321305.1 NP_001308234.1 4 2012
NM_152392.4 NP_689605.1 4 2012
XM_011532541.2 XP_011530843.1 5 970
XM_011532543.2 XP_011530845.1 6 1254
XM_011532545.2 XP_011530847.1 5 970
XM_017003377.1 XP_016858866.1 5 970
XM_017003378.1 XP_016858867.1 6 929
GenBank mRNA AF542548.1 - 3 213
AK092505.1 - 4 399
AK092817.1 - 4 1416
AL833813.1 - 4 1747
BC063789.1 - 4 1901
BC113671.1 - 4 1009 8322723
BC117278.1 - 4 1009 40125829

View Full Product Details