Gene Transcripts

Gene Symbol
A2ML1
Entrez Gene ID
144568
Gene Name
alpha-2-macroglobulin like 1
Gene Aliases
CPAMD9
Chromosome Location
Chr. 12: 8822472 - 8887202 on Build GRCh38
UniGene ID
Hs.620532
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282424.2 NP_001269353.1 21 2805
NM_144670.5 NP_653271.2 32 4290
XM_011520566.2 XP_011518868.1 32 4247
XM_011520567.2 XP_011518869.1 32 4247
XM_017018868.1 XP_016874357.1 32 4244
XM_017018869.1 XP_016874358.1 32 4244
XM_017018870.1 XP_016874359.1 32 4208
GenBank mRNA AK057908.1 - 3 292
AK096448.1 - 10 1279
AK122624.1 - 26 3376
AK123591.1 - 15 1943
AK123592.1 - 21 2760
AK123601.1 - 29 3667
AK290355.1 - 32 4143
AK302555.1 - 21 2805
AL832139.1 - 32 4141
AL832750.1 - 10 1015
AL833479.1 - 30 3806
BC093840.1 - 3 243
BC112131.1 - 3 243
BX647674.1 - 14 1745
BX648231.1 - 33 4142

View Full Product Details