Gene Transcripts

Gene Symbol
ANKRD29
Entrez Gene ID
147463
Gene Name
ankyrin repeat domain 29
Gene Aliases
Chromosome Location
Chr. 18: 23598926 - 23662914 on Build GRCh38
UniGene ID
Hs.355689
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308238.1 NP_001295167.1 4 429
NM_173505.3 NP_775776.2 4 429
XM_011525831.2 XP_011524133.1 3 2369
XM_011525832.2 XP_011524134.1 3 2487
GenBank mRNA AK057782.1 - 4 359
AK313667.1 - 4 347
BC030622.1 - 4 434 4818619

View Full Product Details