Gene Transcripts
- Gene Symbol
- AIFM3
- Entrez Gene ID
- 150209
- Gene Name
- apoptosis inducing factor, mitochondria associated 3
- Gene Aliases
- AIFL
- Chromosome Location
- Chr. 22: 20965130 - 20981360 on Build GRCh38
- UniGene ID
- Hs.723246
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001018060.2 |
NP_001018070.1
|
15, 16 |
1567 |
|
NM_001146288.1 |
NP_001139760.1
|
15, 16 |
1652 |
|
NM_144704.2 |
NP_653305.1
|
15, 16 |
1567 |
|
NR_027464.1 |
|
14, 15 |
1524 |
|
GenBank mRNA |
AK055035.1 |
- |
15, 16 |
1551 |
|
AK094844.1 |
- |
15, 16 |
1556 |
|
AK126917.1 |
- |
1, 2 |
894 |
|
AK295581.1 |
- |
14, 15 |
1427 |
|
AK316042.1 |
- |
14, 15 |
1420 |
|
AY927594.1 |
- |
4, 5 |
339 |
|
BC032485.1 |
- |
15, 16 |
1534 |
5221203 |
CR456342.1 |
- |
14, 15 |
1386 |
|
View Full Product Details