Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040202.1 | NP_001035292.1 | 2 | 467 | |
XM_005262769.4 | XP_005262826.1 | 2 | 467 | ||
XM_017007785.1 | XP_016863274.1 | 2 | 467 | ||
GenBank mRNA | AK055774.1 | - | 2 | 255 | |
AK291232.1 | - | 2 | 447 | ||
AX775925.1 | - | 2 | 461 | ||
AY424281.1 | - | 2 | 394 | ||
BC031256.1 | - | 2 | 433 | 5271599 | |
BC047510.1 | - | 2 | 442 | 5297045 | |
DA410845.1 | - | 2 | 467 |